Assay Details
Target Gene Details
Entrez Gene ID: | 159963 |
Gene Name: | solute carrier family 5 member 12 |
Gene Aliases: |
SMCT2 |
Location: |
Chr.11:26667019-26723356 on Build GRCh38 |
Assay Gene Location: | Within Intron 16 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SLC5A12 | NM_178498.3 | NP_848593.2 | ||
| XM_006718155.3 | XP_006718218.1 | |||
| XM_011519920.2 | XP_011518222.1 | |||
| XM_017017244.1 | XP_016872733.1 | |||
| AY299482.1 | ||||
| BC041454.1 | AAH41454.1 | |||
| CR747563.1 |
More Information
Set Membership: |
|

Genomic Map