Assay Details
Target Gene Details
Entrez Gene ID: | 80150 |
Gene Name: | asparaginase like 1 |
Gene Aliases: |
ALP, ALP1, CRASH |
Location: |
Chr.11:62337302-62401399 on Build GRCh38 |
Assay Gene Location: | Within Intron 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ASRGL1 | XM_011545265.2 | XP_011543567.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1039105 | Chr.11:62356434 - 62404820 on Build GRCh38 | Gain |
|
| nsv468586 | Chr.11:62372597 - 62415535 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map