Assay Details
Target Gene Details
Entrez Gene ID: | 113130 |
Gene Name: | cell division cycle associated 5 |
Gene Aliases: |
SORORIN |
Location: |
Chr.11:65061009-65084143 on Build GRCh38 |
Assay Gene Location: | Within Intron 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CDCA5 | XM_005273733.3 | XP_005273790.1 | ||
| XM_005273734.3 | XP_005273791.1 | |||
| XM_011544743.2 | XP_011543045.1 | |||
| XM_011544747.2 | XP_011543049.1 | |||
| XM_011544748.2 | XP_011543050.1 | |||
| XM_011544749.2 | XP_011543051.1 | |||
| XM_017017158.1 | XP_016872647.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv526181 | Chr.11:64870569 - 65489858 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs115676868] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map