Assay Details
Target Gene Details
Entrez Gene ID: | 7275 |
Gene Name: | tubby bipartite transcription factor |
Gene Aliases: |
RDOB, rd5 |
Location: |
Chr.11:8019180-8106107 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TUB | NM_003320.4 | NP_003311.2 | ||
| XM_005253109.3 | XP_005253166.1 | |||
| AK293528.1 | ||||
| BC075031.2 | AAH75031.1 | |||
| BC075032.2 | AAH75032.1 | |||
| U82467.1 | AAB53699.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv428249 | Chr.11:8047047 - 8194822 on Build GRCh38 | Loss |
|
| esv1001231 | Chr.11:8050228 - 8050991 on Build GRCh38 | Insertion |
|
More Information
Additional Information:
For this assay, SNP(s) [rs115695740] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map