Assay Details
Target Gene Details
Entrez Gene ID: | 80150 |
Gene Name: | asparaginase like 1 |
Gene Aliases: |
ALP, ALP1, CRASH |
Location: |
Chr.11:62337302-62401399 on Build GRCh38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ASRGL1 | NM_001083926.1 | 7 | 2228 | NP_001077395.1 |
| NM_025080.3 | 7 | 2151 | NP_079356.3 | |
| XM_005274305.3 | 6 | 1868 | XP_005274362.1 | |
| XM_005274306.2 | 6 | 1930 | XP_005274363.1 | |
| XM_011545265.2 | XP_011543567.1 | |||
| AK090908.1 | 7 | 2000 | ||
| AK091894.1 | 7 | 2082 | ||
| BC006267.2 | 4 | 1541 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1039105 | Chr.11:62356434 - 62404820 on Build GRCh38 | Gain |
|
| nsv468586 | Chr.11:62372597 - 62415535 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map