Assay Details
Target Gene Details
Entrez Gene ID: | 85458 |
Gene Name: | DIX domain containing 1 |
Gene Aliases: |
CCD1 |
Location: |
Chr.11:111927144-112022650 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 2 - Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DIXDC1 | NM_001278542.1 | NP_001265471.1 | ||
| XM_017018466.1 | XP_016873955.1 | |||
| XM_017018467.1 | XP_016873956.1 | |||
| BC048294.1 | AAH48294.1 | |||
| DB459100.1 | ||||
| HY037299.1 |
More Information
Set Membership: |
|

Genomic Map