Assay Details
Target Gene Details
Entrez Gene ID: | 9873 |
Gene Name: | FCH and double SH3 domains 2 |
Gene Aliases: |
NWK, SH3MD3 |
Location: |
Chr.11:72836743-73142682 on Build GRCh38 |
Assay Gene Location: | Within Intron 15 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FCHSD2 | NM_014824.2 | NP_055639.2 | ||
| XM_011545409.1 | XP_011543711.1 | |||
| XM_011545410.2 | XP_011543712.1 | |||
| XM_017018632.1 | XP_016874121.1 | |||
| AB018312.1 | BAA34489.2 | |||
| AK296376.1 | ||||
| AK297928.1 | ||||
| AL133567.1 | CAB63720.1 | |||
| BC010394.1 | AAH10394.1 | |||
| BC017751.1 | AAH17751.1 |
More Information
Additional Information:
For this assay, SNP(s) [rs78849193] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map