Assay Details
Target Gene Details
Entrez Gene ID: | 5962 |
Gene Name: | radixin |
Gene Aliases: |
DFNB24 |
Location: |
Chr.11:110174880-110296712 on Build GRCh38 |
Assay Gene Location: | Within Intron 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RDX | NM_001260492.1 | NP_001247421.1 | ||
| NM_001260493.1 | NP_001247422.1 | |||
| NM_001260495.1 | NP_001247424.1 | |||
| NM_001260496.1 | NP_001247425.1 | |||
| DQ916738.1 | ||||
| DQ916739.1 | ||||
| DQ916740.1 | ||||
| DQ916741.1 | ||||
| DQ916742.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv832265 | Chr.11:110041489 - 110225405 on Build GRCh38 | Gain |
|
| nsv1152468 | Chr.11:110208174 - 110220275 on Build GRCh38 | Duplication |
|
More Information
Additional Information:
For this assay, SNP(s) [rs75430240] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map