Assay Details
Target Gene Details
Entrez Gene ID: | 101928200 |
Gene Name: | MRGPRF antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.11:69012354-69018447 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MRGPRF-AS1 | NR_120541.1 | 3 | 411 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv555299 | Chr.11:69002565 - 69053408 on Build GRCh38 | Loss |
|
| nsv832196 | Chr.11:68995114 - 69165820 on Build GRCh38 | Loss |
|

Genomic Map