Assay Details
Target Gene Details
Entrez Gene ID: | 867 |
Gene Name: | Cbl proto-oncogene |
Gene Aliases: |
C-CBL, CBL2, FRA11B, NSLL, RNF55 |
Location: |
Chr.11:119206276-119308149 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CBL | NM_005188.3 | NP_005179.2 | ||
| XM_011543057.2 | XP_011541359.1 | |||
| BC132733.1 | ||||
| BC136463.1 | ||||
| BG722940.1 | ||||
| DB457271.2 | ||||
| X57110.1 | CAA40393.1 |
More Information
Set Membership: |
|

Genomic Map