Assay Details
Target Gene Details
Entrez Gene ID: | 3746 |
Gene Name: | potassium voltage-gated channel subfamily C member 1 |
Gene Aliases: |
EPM7, KV3.1, KV4, NGK2 |
Location: |
Chr.11:17734810-17783055 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KCNC1 | NM_001112741.1 | NP_001106212.1 | ||
| NM_004976.4 | NP_004967.1 | |||
| XM_011520080.2 | XP_011518382.1 | |||
| BC107129.2 | AAI07130.1 | |||
| DN993192.1 | ||||
| L00621.1 | ||||
| S56770.1 | AAB25764.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv553589 | Chr.11:17723445 - 17788177 on Build GRCh38 | Loss |
|
| nsv553588 | Chr.11:17691960 - 17770275 on Build GRCh38 | Loss |
|
| nsv553567 | Chr.11:16890704 - 17946708 on Build GRCh38 | Gain |
|
| nsv1038119 | Chr.11:17723368 - 17773030 on Build GRCh38 | Gain |
|
| nsv521511 | Chr.11:17727481 - 17749940 on Build GRCh38 | Loss |
|
| nsv509390 | Chr.11:17726193 - 17855296 on Build GRCh38 | Insertion |
|
| nsv825772 | Chr.11:17701983 - 17761581 on Build GRCh38 | Gain |
|
| nsv516767 | Chr.11:17737040 - 17788177 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs74636657] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map