Assay Details
Target Gene Details
Entrez Gene ID: | 2348 |
Gene Name: | folate receptor 1 |
Gene Aliases: |
FBP, FOLR |
Location: |
Chr.11:72189558-72196323 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FOLR1 | NM_000802.3 | NP_000793.1 | ||
| NM_016724.2 | NP_057936.1 | |||
| NM_016725.2 | NP_057937.1 | |||
| NM_016729.2 | NP_057941.1 | |||
| AK223527.1 | BAD97247.1 | |||
| BC002947.2 | AAH02947.1 | |||
| BT007158.1 | AAP35822.1 | |||
| CR542019.1 | CAG46816.1 | |||
| J05013.1 | AAA35823.1 | |||
| KU257686.1 | ||||
| M25317.1 | AAA74896.1 | |||
| M28099.1 | AAA35822.1 | |||
| X62753.1 | CAA44610.1 |
More Information
Additional Information:
For this assay, SNP(s) [rs76101020] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map