Assay Details
Target Gene Details
Entrez Gene ID: | 3995 |
Gene Name: | fatty acid desaturase 3 |
Gene Aliases: |
CYB5RP, LLCDL3 |
Location: |
Chr.11:61873523-61892224 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FADS3 | NM_021727.4 | NP_068373.1 | ||
| XM_011545023.2 | XP_011543325.1 | |||
| XM_017017723.1 | XP_016873212.1 | |||
| XM_017017724.1 | XP_016873213.1 | |||
| AF084560.1 | AAG23122.1 | |||
| AF134404.1 | AAD31282.1 | |||
| AK295473.1 | ||||
| AK310523.1 | ||||
| BC004901.2 | AAH04901.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3626586 | Chr.11:61881703 - 61885743 on Build GRCh38 | Loss |
|
| dgv217e199 | Chr.11:61881645 - 61885775 on Build GRCh38 | Deletion |
|
| nsv528697 | Chr.11:61690385 - 61919561 on Build GRCh38 | Loss |
|
| nsv468585 | Chr.11:61848540 - 61897520 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs74626285] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map