Assay Details
Target Gene Details
Entrez Gene ID: | 5241 |
Gene Name: | progesterone receptor |
Gene Aliases: |
NR3C3, PR |
Location: |
Chr.11:101029624-101130681 on Build GRCh38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PGR | NM_000926.4 | 8 | 12898 | NP_000917.3 |
| NM_001202474.3 | 8 | 12148 | NP_001189403.1 | |
| NM_001271161.2 | 7 | 11842 | NP_001258090.1 | |
| NM_001271162.1 | 8 | 10818 | NP_001258091.1 | |
| NR_073141.2 | 8 | 12096 | ||
| NR_073142.2 | 7 | 11979 | ||
| NR_073143.2 | 6 | 11711 | ||
| AL137566.1 | 1 | 3461 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1159933 | Chr.11:101014626 - 101057558 on Build GRCh38 | Duplication |
|
More Information
Set Membership: |
|

Genomic Map