Assay Details
Target Gene Details
Entrez Gene ID: | 338645 |
Gene Name: | leucine zipper protein 2 |
Gene Aliases: |
KFSP2566, PRO6246 |
Location: |
Chr.11:24494304-25082640 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LUZP2 | XM_011520056.2 | 1 | 2261 | XP_011518358.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv553819 | Chr.11:24477824 - 24523807 on Build GRCh38 | Loss |
|
| nsv1040177 | Chr.11:23010756 - 24853532 on Build GRCh38 | Gain |
|
| nsv467755 | Chr.11:24385835 - 24506412 on Build GRCh38 | Loss |
|
| nsv469945 | Chr.11:24493977 - 24523807 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs80015235] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map