Assay Details
Target Gene Details
Entrez Gene ID: | 101929538 |
Gene Name: | uncharacterized LOC101929538 |
Gene Aliases: |
- |
Location: |
Chr.11:128681240-128686922 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC101929538 | NR_135072.1 | |||
| AK093020.1 | ||||
| DA141900.1 |

Genomic Map