Assay Details
Target Gene Details
Entrez Gene ID: | 4925 |
Gene Name: | nucleobindin 2 |
Gene Aliases: |
HEL-S-109, NEFA |
Location: |
Chr.11:17260340-17331881 on Build GRCh38 |
Assay Gene Location: | Within Intron 7 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv524611 | Chr.11:17279297 - 17294482 on Build GRCh38 | Loss |
|
| nsv7692 | Chr.11:17266058 - 17299821 on Build GRCh38 | Insertion |
|
| nsv553567 | Chr.11:16890704 - 17946708 on Build GRCh38 | Gain |
|
| nsv467711 | Chr.11:17006008 - 17299250 on Build GRCh38 | Gain |
|
| nsv469762 | Chr.11:17149143 - 17299144 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map