Assay Details
Target Gene Details
Entrez Gene ID: | 3312 |
Gene Name: | heat shock protein family A (Hsp70) member 8 |
Gene Aliases: |
HEL-33, HEL-S-72p, HSC54, HSC70, HSC71, HSP71, HSP73, HSPA10, LAP-1, LAP1, NIP71 |
Location: |
Chr.11:123057492-123062366 on Build GRCh38 |
Assay Gene Location: | Within Intron 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| HSPA8 | NM_006597.5 | NP_006588.1 | ||
| NM_153201.3 | NP_694881.1 | |||
| XM_011542798.1 | XP_011541100.1 | |||
| AB034951.1 | BAB18615.1 | |||
| AB073886.1 | ||||
| AF217511.1 | AAF67622.1 | |||
| AF352832.1 | AAK17898.1 | |||
| AK000571.1 | BAA91262.1 | |||
| AK096100.1 | ||||
| AK129885.1 | ||||
| AK222628.1 | BAD96348.1 | |||
| AK222785.1 | BAD96505.1 | |||
| AK292067.1 | ||||
| AK303935.1 | ||||
| BC007276.1 | AAH07276.2 | |||
| BC008907.2 | AAH08907.2 | |||
| BC015699.1 | AAH15699.1 | |||
| BC016179.1 | AAH16179.1 | |||
| BC016660.1 | AAH16660.1 | |||
| BC019816.2 | AAH19816.1 | |||
| EU668349.1 | ||||
| FJ224294.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv983069 | Chr.11:123057251 - 123060217 on Build GRCh38 | Duplication |
|
| nsv1039284 | Chr.11:122985952 - 123083303 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map