Assay Details
Target Gene Details
Entrez Gene ID: | 738 |
Gene Name: | VPS51, GARP complex subunit |
Gene Aliases: |
ANG2, ANG3, C11orf2, C11orf3, FFR |
Location: |
Chr.11:65096115-65111860 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| VPS51 | NM_013265.3 | NP_037397.2 | ||
| NR_073519.1 | ||||
| AF024631.2 | AAF21627.2 | |||
| BC007198.1 | AAH07198.1 | |||
| BC010540.1 | AAH10540.1 | |||
| BC017438.1 | AAH17438.1 | |||
| BG391473.1 | ||||
| BI752026.1 | ||||
| BI823833.1 | ||||
| BI828867.1 | ||||
| BX401315.2 | ||||
| CD388131.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv526181 | Chr.11:64870569 - 65489858 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map