Assay Details
Target Gene Details
Entrez Gene ID: | 10335 |
Gene Name: | murine retrovirus integration site 1 homolog |
Gene Aliases: |
IRAG, JAW1L |
Location: |
Chr.11:10573091-10693988 on Build GRCh38 |
Assay Gene Location: | Within Intron 15 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MRVI1 | NM_001098579.2 | NP_001092049.2 | ||
| NM_001100163.2 | NP_001093633.1 | |||
| NM_001100167.2 | NP_001093637.1 | |||
| NM_001206880.1 | NP_001193809.1 | |||
| NM_001206881.1 | NP_001193810.1 | |||
| NM_130385.3 | NP_569056.4 | |||
| AF081249.1 | AAD25922.1 | |||
| AF081250.1 | AAD25923.1 | |||
| AK127209.1 | ||||
| AK296336.1 | ||||
| AK300358.1 | ||||
| AK301296.1 | ||||
| AK304702.1 | ||||
| BC117127.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv832063 | Chr.11:10596938 - 10763734 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map