Assay Details
Target Gene Details
Entrez Gene ID: | 7704 |
Gene Name: | zinc finger and BTB domain containing 16 |
Gene Aliases: |
PLZF, ZNF145 |
Location: |
Chr.11:114059576-114256765 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ZBTB16 | NM_001018011.1 | NP_001018011.1 | ||
| NM_006006.4 | NP_005997.2 | |||
| XM_005271658.4 | XP_005271715.1 | |||
| XM_017018257.1 | XP_016873746.1 | |||
| XM_017018258.1 | XP_016873747.1 | |||
| XM_017018259.1 | XP_016873748.1 | |||
| AB208916.1 | BAD92153.1 | |||
| BC026902.1 | AAH26902.1 | |||
| BC029812.1 | AAH29812.1 | |||
| BX648973.1 | ||||
| KU178400.1 | ||||
| KU178401.1 | ||||
| Z19002.1 | CAA79489.1 |
More Information
Additional Information:
For this assay, SNP(s) [rs118050126] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map