Assay Details
Target Gene Details
Entrez Gene ID: | 144486 |
Gene Name: | CEP83 antisense RNA 1 (head to head) |
Gene Aliases: |
CCDC41-AS1 |
Location: |
Chr.12:94460003-94462570 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CEP83-AS1 | NR_027035.1 | 1 | 51 | |
| AK057269.1 | 1 | 51 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv832490 | Chr.12:94330721 - 94496512 on Build GRCh38 | Gain |
|
| nsv516550 | Chr.12:94338818 - 94496272 on Build GRCh38 | Loss |
|

Genomic Map