Assay Details
Target Gene Details
Entrez Gene ID: | 5074 |
Gene Name: | pro-apoptotic WT1 regulator |
Gene Aliases: |
PAR4, Par-4 |
Location: |
Chr.12:79587791-79691097 on Build GRCh38 |
Assay Gene Location: | Within Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PAWR | XM_006719435.2 | 7 | 3136 | XP_006719498.1 |
| XM_017019377.1 | 7 | 3040 | XP_016874866.1 |
More Information
Set Membership: |
|

Genomic Map