Assay Details
Target Gene Details
Entrez Gene ID: | 4633 |
Gene Name: | myosin light chain 2 |
Gene Aliases: |
CMH10, MLC-2s/v, MLC2 |
Location: |
Chr.12:110910819-110920600 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MYL2 | NM_000432.3 | NP_000423.2 | ||
| AF020768.1 | AAB91993.1 | |||
| AK312013.1 | ||||
| BC015821.1 | AAH15821.1 | |||
| BC031006.1 | AAH31006.1 | |||
| BC031008.1 | AAH31008.1 | |||
| BX538165.1 | ||||
| CR456962.1 | CAG33243.1 | |||
| CR541957.1 | CAG46755.1 | |||
| M22815.1 | AAA91832.1 | |||
| S69022.1 | AAB29658.2 | |||
| X14332.1 | CAA32510.1 | |||
| X57542.1 | CAA40761.1 | |||
| X66141.1 | CAA46931.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3630744 | Chr.12:110849055 - 110980384 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map