Assay Details
Target Gene Details
Entrez Gene ID: | 55785 |
Gene Name: | FYVE, RhoGEF and PH domain containing 6 |
Gene Aliases: |
ZFYVE24 |
Location: |
Chr.12:95076749-95217464 on Build GRCh38 |
Assay Gene Location: | Within Exon 21 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FGD6 | NM_018351.3 | 21 | 8549 | NP_060821.3 |
| AK026881.1 | 1 | 963 |
More Information
Set Membership: |
|

Genomic Map