Assay Details
Target Gene Details
Entrez Gene ID: | 196383 |
Gene Name: | Rab interacting lysosomal protein like 2 |
Gene Aliases: |
RLP2 |
Location: |
Chr.12:123408567-123436733 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RILPL2 | XM_011538012.2 | XP_011536314.1 | ||
| XM_017018959.1 | 4 | 1927 | XP_016874448.1 |
More Information
Set Membership: |
|

Genomic Map