Assay Details
Target Gene Details
Entrez Gene ID: | 3375 |
Gene Name: | islet amyloid polypeptide |
Gene Aliases: |
DAP, IAP |
Location: |
Chr.12:21354928-21379980 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| IAPP | XM_017019253.1 | XP_016874742.1 | ||
| AA429852.1 | ||||
| AA446156.1 |
Target Gene Details
Entrez Gene ID: | 6579 |
Gene Name: | solute carrier organic anion transporter family member 1A2 |
Gene Aliases: |
OATP, OATP-A, OATP1A2, SLC21A3 |
Location: |
Chr.12:21264600-21403657 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SLCO1A2 | NM_134431.3 | NP_602307.1 | ||
| XM_011520819.1 | XP_011519121.1 | |||
| AF279784.1 | AAG30037.1 | |||
| AK316249.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv557735 | Chr.12:21290348 - 21386023 on Build GRCh38 | Loss |
|
| nsv557737 | Chr.12:21354768 - 21402757 on Build GRCh38 | Loss |
|
| esv3628806 | Chr.12:21343905 - 21380714 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map