Assay Details
Target Gene Details
Entrez Gene ID: | 88455 |
Gene Name: | ankyrin repeat domain 13A |
Gene Aliases: |
ANKRD13, NY-REN-25 |
Location: |
Chr.12:109999169-110039432 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ANKRD13A | NM_033121.1 | NP_149112.1 | ||
| XM_005253980.2 | XP_005254037.1 | |||
| XM_005253981.3 | XP_005254038.1 | |||
| XM_005253982.3 | XP_005254039.1 | |||
| XM_005253984.1 | XP_005254041.1 | |||
| XM_011538937.1 | XP_011537239.1 | |||
| XM_011538938.2 | XP_011537240.1 | |||
| XM_017020157.1 | XP_016875646.1 | |||
| XM_017020158.1 | XP_016875647.1 | |||
| XM_017020159.1 | XP_016875648.1 | |||
| XM_017020160.1 | XP_016875649.1 | |||
| XM_017020161.1 | XP_016875650.1 | |||
| XM_017020162.1 | XP_016875651.1 | |||
| XM_017020163.1 | XP_016875652.1 | |||
| AF064604.1 | AAC17109.1 | |||
| AK025261.1 | ||||
| AK302535.1 | ||||
| AK308923.1 | ||||
| AY364251.1 | AAQ76810.1 | |||
| BC032833.2 | AAH32833.2 |

Genomic Map