Assay Details
Target Gene Details
Entrez Gene ID: | 3824 |
Gene Name: | killer cell lectin like receptor D1 |
Gene Aliases: |
CD94 |
Location: |
Chr.12:10238385-10329607 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KLRD1 | XM_017019290.1 | XP_016874779.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1054811 | Chr.12:10197988 - 10287406 on Build GRCh38 | Loss |
|
| nsv832328 | Chr.12:10245472 - 10467844 on Build GRCh38 | Gain |
|
| nsv557454 | Chr.12:10264661 - 10288286 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map