Assay Details
Target Gene Details
Entrez Gene ID: | 121278 |
Gene Name: | tryptophan hydroxylase 2 |
Gene Aliases: |
ADHD7, NTPH |
Location: |
Chr.12:71938846-72032441 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TPH2 | NM_173353.3 | NP_775489.2 | ||
| AK094614.1 | BAC04385.1 | |||
| AY098914.1 | AAM28946.1 | |||
| BC114442.1 | ||||
| BC114499.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv518498 | Chr.12:71913836 - 72114999 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map