Assay Details
Target Gene Details
Entrez Gene ID: | 26628 |
Gene Name: | olfactory receptor family 7 subfamily E member 47 pseudogene |
Gene Aliases: |
OR7E141 |
Location: |
Chr.12:52079696-52108319 on Build GRCh38 |
Assay Gene Location: | Within Intron 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| OR7E47P | NR_120437.1 | |||
| NR_120439.1 | ||||
| NR_120440.1 | ||||
| AA229829.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv470292 | Chr.12:52066640 - 52127560 on Build GRCh38 | Loss |
|
| nsv832409 | Chr.12:52055382 - 52214067 on Build GRCh38 | Loss |
|
| nsv983324 | Chr.12:52094618 - 52108250 on Build GRCh38 | Duplication |
|
| nsv558898 | Chr.12:52009326 - 52122725 on Build GRCh38 | Loss |
|

Genomic Map