Assay Details
Target Gene Details
Entrez Gene ID: | 65985 |
Gene Name: | acetoacetyl-CoA synthetase |
Gene Aliases: |
ACSF1, SUR-5 |
Location: |
Chr.12:125065367-125143333 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| AACS | NM_001319839.1 | NP_001306768.1 | ||
| NM_001319840.1 | NP_001306769.1 | |||
| NM_023928.4 | NP_076417.2 | |||
| XM_011538692.2 | XP_011536994.1 | |||
| AB054121.1 | BAD22560.1 | |||
| AK022451.1 | BAB14040.1 | |||
| AK022740.1 | BAB14216.1 | |||
| AK024036.1 | BAB14793.1 | |||
| AK304176.1 | ||||
| AK308779.1 | ||||
| AL833047.1 | CAH56309.1 | |||
| AY091468.1 | AAM44124.1 | |||
| BC040490.1 | AAH40490.2 | |||
| BC041000.1 | AAH41000.1 | |||
| BC051862.1 | AAH51862.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1051020 | Chr.12:125015140 - 125809096 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map