Assay Details
Target Gene Details
Entrez Gene ID: | 51699 |
Gene Name: | VPS29, retromer complex component |
Gene Aliases: |
DC15, DC7, PEP11 |
Location: |
Chr.12:110491523-110502140 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| VPS29 | NM_001282150.1 | NP_001269079.1 | ||
| NM_001282151.1 | NP_001269080.1 | |||
| NM_016226.4 | NP_057310.1 | |||
| NM_057180.2 | NP_476528.1 | |||
| NR_104099.1 | ||||
| NR_104100.1 | ||||
| NR_104101.1 | ||||
| NR_104102.1 | ||||
| XM_006719459.3 | XP_006719522.1 | |||
| XM_006719460.3 | XP_006719523.1 | |||
| XM_011538484.2 | XP_011536786.1 | |||
| XM_017019457.1 | XP_016874946.1 | |||
| XM_017019459.1 | XP_016874948.1 | |||
| AA148059.1 | ||||
| AB578891.1 | ||||
| AB578892.1 | ||||
| AB618733.1 | ||||
| AF168716.1 | AAF87318.1 | |||
| AF175264.1 | AAF89952.1 | |||
| AF193795.1 | AAF04596.1 | |||
| AF201946.1 | AAF17238.1 | |||
| AK129767.1 | ||||
| AL136614.1 | CAB66549.1 | |||
| AL832866.1 | 1 | 1363 | CAI46196.1 | |
| BC000880.1 | AAH00880.1 | |||
| BC015095.1 | ||||
| BC095446.1 | AAH95446.1 | |||
| BG707325.1 | ||||
| BM806646.1 | ||||
| BU568689.1 | ||||
| CB158821.1 |
More Information
Set Membership: |
|

Genomic Map