Assay Details
Target Gene Details
Entrez Gene ID: | 613227 |
Gene Name: | HIG1 hypoxia inducible domain family member 1C |
Gene Aliases: |
GM921 |
Location: |
Chr.12:50936223-50972592 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| HIGD1C | XM_017019783.1 | 1 | 602 | XP_016875272.1 |
| XM_017019784.1 | 1 | 602 | XP_016875273.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv34039 | Chr.12:50932919 - 51278127 on Build GRCh38 | Loss |
|

Genomic Map