Assay Details
Target Gene Details
Entrez Gene ID: | 51134 |
Gene Name: | centrosomal protein 83 |
Gene Aliases: |
CCDC41, NPHP18, NY-REN-58 |
Location: |
Chr.12:94268832-94459991 on Build GRCh38 |
Assay Gene Location: |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CEP83 |
Target Gene Details
Entrez Gene ID: | 10154 |
Gene Name: | plexin C1 |
Gene Aliases: |
CD232, PLXN-C1, VESPR |
Location: |
Chr.12:94148696-94307675 on Build GRCh38 |
Assay Gene Location: | Within Intron 23 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PLXNC1 | NM_005761.2 | NP_005752.1 | ||
| NR_037687.1 | ||||
| XM_006719186.3 | XP_006719249.1 | |||
| XM_011537730.2 | XP_011536032.1 | |||
| XM_017018671.1 | XP_016874160.1 | |||
| XM_017018672.1 | XP_016874161.1 | |||
| AB208934.1 | BAD92171.1 | |||
| AF030339.1 | AAC18823.1 | |||
| AK295226.1 | ||||
| AK304327.1 | ||||
| AK307920.1 | ||||
| AK308985.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv520739 | Chr.12:94271406 - 94276428 on Build GRCh38 | Loss |
|
| nsv832489 | Chr.12:94186485 - 94352982 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map