Assay Details
Target Gene Details
Entrez Gene ID: | 101928002 |
Gene Name: | uncharacterized LOC101928002 |
Gene Aliases: |
- |
Location: |
Chr.12:69713633-69738568 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC101928002 | NR_110072.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3423810 | Chr.12:69728828 - 69728861 on Build GRCh38 | Insertion |
|
| esv3304014 | Chr.12:69728834 - 69728835 on Build GRCh38 | Mobile Element Insertion |
|

Genomic Map