Assay Details
Target Gene Details
Entrez Gene ID: | 92558 |
Gene Name: | BICD family like cargo adaptor 1 |
Gene Aliases: |
BICDR-1, BICDR1, CCDC64, H_267D11.1 |
Location: |
Chr.12:119989806-120094495 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| BICDL1 | NM_207311.2 | NP_997194.2 | ||
| XM_005253991.4 | XP_005254048.1 | |||
| XM_006719694.3 | XP_006719757.1 | |||
| XM_006719696.3 | XP_006719759.1 | |||
| XM_006719697.3 | XP_006719760.1 | |||
| XM_011538999.2 | XP_011537301.1 | |||
| XM_011539000.2 | XP_011537302.1 | |||
| AK129960.1 | BAC85259.1 | |||
| U88834.1 |
More Information
Additional Information:
For this assay, SNP(s) [rs77047679] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map