Assay Details
Target Gene Details
Entrez Gene ID: | 192111 |
Gene Name: | PGAM family member 5, mitochondrial serine/threonine protein phosphatase |
Gene Aliases: |
BXLBV68 |
Location: |
Chr.12:132710807-132722737 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PGAM5 | NM_001170543.1 | NP_001164014.1 | ||
| NM_001170544.1 | NP_001164015.1 | |||
| NM_138575.3 | NP_612642.2 | |||
| AK097688.1 | ||||
| BC008196.1 | AAH08196.1 | |||
| DR155760.1 | ||||
| EU249757.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1047500 | Chr.12:132666737 - 132763400 on Build GRCh38 | Gain |
|
| nsv1037074 | Chr.12:132613063 - 132801953 on Build GRCh38 | Gain |
|
| esv3631311 | Chr.12:132713141 - 132737944 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map