Assay Details
Target Gene Details
Entrez Gene ID: | 11016 |
Gene Name: | activating transcription factor 7 |
Gene Aliases: |
ATFA |
Location: |
Chr.12:53507856-53626415 on Build GRCh38 |
Assay Gene Location: | Within Exon 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ATF7 | NM_001130060.1 | 12 | 5318 | NP_001123532.1 |
| NM_006856.2 | 12 | 5381 | NP_006847.1 | |
| NR_073163.1 | ||||
| XM_005268587.3 | 12 | 5333 | XP_005268644.1 | |
| XM_017018722.1 | 12 | 6521 | XP_016874211.1 | |
| AK298853.1 | ||||
| DB047160.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv983486 | Chr.12:53503642 - 53514027 on Build GRCh38 | Duplication |
|
More Information
Set Membership: |
|

Genomic Map