Assay Details
Target Gene Details
Entrez Gene ID: | 5463 |
Gene Name: | POU class 6 homeobox 1 |
Gene Aliases: |
BRN5, MPOU, TCFB1 |
Location: |
Chr.12:51186936-51218134 on Build GRCh38 |
Assay Gene Location: | Within Intron 7 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2745882 | Chr.12:51199208 - 51199283 on Build GRCh38 | Deletion |
|
| esv34039 | Chr.12:50932919 - 51278127 on Build GRCh38 | Loss |
|
| nsv558876 | Chr.12:51197584 - 51218946 on Build GRCh38 | Gain |
|
| esv2745880 | Chr.12:51198884 - 51199408 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map