Assay Details
Target Gene Details
Entrez Gene ID: | 80212 |
Gene Name: | coiled-coil domain containing 92 |
Gene Aliases: |
- |
Location: |
Chr.12:123936409-123972985 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CCDC92 | NM_001304957.1 | NP_001291886.1 | ||
| NM_001304958.1 | NP_001291887.1 | |||
| NM_001304959.1 | NP_001291888.1 | |||
| NM_001304960.1 | NP_001291889.1 | |||
| NM_001304961.1 | NP_001291890.1 | |||
| NM_025140.2 | NP_079416.1 | |||
| AK026124.1 | BAB15366.1 | |||
| AK222661.1 | BAD96381.1 | |||
| AW953473.1 | ||||
| BC017186.2 | AAH17186.1 | |||
| BX394686.2 | ||||
| DA263704.1 | ||||
| DA370549.1 | ||||
| HY035895.1 |
More Information
Additional Information:
For this assay, SNP(s) [rs112619493] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map