Assay Details
Target Gene Details
Entrez Gene ID: | 100526761 |
Gene Name: | CCDC169-SOHLH2 readthrough |
Gene Aliases: |
C13orf38-SOHLH2 |
Location: |
Chr.13:36168208-36297855 on Build GRCh38 |
Assay Gene Location: | Within Intron 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CCDC169-SOHLH2 | NM_001198910.1 | NP_001185839.1 | ||
| AK301863.1 |
Target Gene Details
Entrez Gene ID: | 54937 |
Gene Name: | spermatogenesis and oogenesis specific basic helix-loop-helix 2 |
Gene Aliases: |
SPATA28, TEB1, bHLHe81 |
Location: |
Chr.13:36168208-36214615 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SOHLH2 | NM_001282147.1 | NP_001269076.1 | ||
| NM_017826.2 | NP_060296.2 | |||
| AK000456.1 | BAA91175.1 | |||
| AY884305.1 | AAW78547.1 | |||
| BC025383.2 | AAH25383.1 | |||
| DB022954.1 |
More Information
Set Membership: |
|

Genomic Map