Assay Details
Target Gene Details
Entrez Gene ID: | 160851 |
Gene Name: | diacylglycerol kinase eta |
Gene Aliases: |
DGKeta |
Location: |
Chr.13:42040036-42256584 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DGKH | NM_001204504.2 | NP_001191433.1 | ||
| NM_152910.5 | NP_690874.2 | |||
| NM_178009.4 | NP_821077.1 | |||
| AB078967.1 | BAC66960.1 | |||
| AB078968.1 | BAC66961.1 | |||
| AK289546.1 | ||||
| DR002010.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2661097 | Chr.13:42068220 - 42073591 on Build GRCh38 | Deletion |
|
| nsv832589 | Chr.13:42062545 - 42203497 on Build GRCh38 | Loss |
|
| esv3631888 | Chr.13:42068228 - 42073595 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map