Assay Details
Target Gene Details
Entrez Gene ID: | 100190939 |
Gene Name: | TPT1 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.13:45341345-45391483 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TPT1-AS1 | NR_024458.1 | |||
| AF318337.1 | AAL55844.1 | |||
| AK124332.1 | BAC85835.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1023 | Chr.13:45368786 - 45391811 on Build GRCh38 | Insertion |
|
| nsv510579 | Chr.13:45305226 - 45424938 on Build GRCh38 | Deletion |
|

Genomic Map