Assay Details
Target Gene Details
Entrez Gene ID: | 9818 |
Gene Name: | nucleoporin 58 |
Gene Aliases: |
NUP45, NUPL1, PRO2463 |
Location: |
Chr.13:25301152-25349795 on Build GRCh38 |
Assay Gene Location: | Within Intron 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NUP58 | NM_001008564.1 | NP_001008564.1 | ||
| NM_014089.3 | NP_054808.1 | |||
| XM_005266600.3 | XP_005266657.1 | |||
| XM_011535328.1 | XP_011533630.1 | |||
| XM_011535329.2 | XP_011533631.1 | |||
| XM_017020880.1 | XP_016876369.1 | |||
| XM_017020881.1 | XP_016876370.1 | |||
| AB007870.1 | BAA23706.2 | |||
| AK123955.1 | ||||
| AK302946.1 | ||||
| AK303409.1 | ||||
| BC001104.2 | AAH01104.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3582237 | Chr.13:25162386 - 25434284 on Build GRCh38 | Gain |
|
| esv2747161 | Chr.13:25031743 - 26209531 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map