Assay Details
Target Gene Details
Entrez Gene ID: | 7278 |
Gene Name: | tubulin alpha 3c |
Gene Aliases: |
TUBA2, bA408E5.3 |
Location: |
Chr.13:19173770-19181852 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TUBA3C | NM_006001.2 | NP_005992.1 | ||
| AA412719.1 | ||||
| BC011721.2 | AAH11721.1 | |||
| DB524026.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1053637 | Chr.13:19125919 - 19231740 on Build GRCh38 | Gain |
|
| esv3631379 | Chr.13:19165899 - 19178877 on Build GRCh38 | Gain |
|
| nsv561202 | Chr.13:19157877 - 19184771 on Build GRCh38 | Loss |
|
| nsv482166 | Chr.13:19173780 - 19181796 on Build GRCh38 | Gain |
|
| nsv523647 | Chr.13:19048003 - 19184659 on Build GRCh38 | Loss |
|
| nsv983548 | Chr.13:19171886 - 19221146 on Build GRCh38 | Duplication |
|
| nsv528089 | Chr.13:19172470 - 19184771 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map