Assay Details
Target Gene Details
Entrez Gene ID: | 55608 |
Gene Name: | ankyrin repeat domain 10 |
Gene Aliases: |
- |
Location: |
Chr.13:110878540-110915107 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ANKRD10 | NM_001286721.1 | NP_001273650.1 | ||
| NM_017664.3 | NP_060134.2 | |||
| NR_104586.1 | ||||
| NR_104587.1 | ||||
| AK000100.1 | BAA90945.1 | |||
| AK095763.1 | ||||
| AK096083.1 | ||||
| BC001727.1 | AAH01727.1 | |||
| BC039715.1 | AAH39715.1 | |||
| BX354283.2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1044340 | Chr.13:110581726 - 111040647 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map