Assay Details
Target Gene Details
Entrez Gene ID: | 103625683 |
Gene Name: | long intergenic non-protein coding RNA 456 |
Gene Aliases: |
- |
Location: |
Chr.13:97172382-97179622 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LINC00456 | NR_125725.1 | 2 | 366 | |
| BG623605.1 | 2 | 366 |
Target Gene Details
Entrez Gene ID: | 10150 |
Gene Name: | muscleblind like splicing regulator 2 |
Gene Aliases: |
MBLL, MBLL39, PRO2032 |
Location: |
Chr.13:97141780-97394120 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MBNL2 | XM_011521045.2 | XP_011519347.1 | ||
| XM_017020308.1 | XP_016875797.1 | |||
| XM_017020309.1 | XP_016875798.1 | |||
| XM_017020310.1 | XP_016875799.1 | |||
| XM_017020311.1 | XP_016875800.1 | |||
| CR749802.1 | CAH18662.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1137 | Chr.13:97151220 - 97196560 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map