Assay Details
Target Gene Details
Entrez Gene ID: | 10161 |
Gene Name: | lysophosphatidic acid receptor 6 |
Gene Aliases: |
ARWH1, HYPT8, LAH3, P2RY5, P2Y5 |
Location: |
Chr.13:48411045-48444704 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LPAR6 | NM_005767.5 | NP_005758.2 | ||
| BC045651.1 | AAH45651.1 |
Target Gene Details
Entrez Gene ID: | 5925 |
Gene Name: | RB transcriptional corepressor 1 |
Gene Aliases: |
OSRC, PPP1R130, RB, p105-Rb, pRb, pp110 |
Location: |
Chr.13:48303747-48481890 on Build GRCh38 |
Assay Gene Location: | Within Intron 18 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RB1 | NM_000321.2 | NP_000312.2 | ||
| XM_011535171.2 | XP_011533473.1 | |||
| AB208788.1 | BAD92025.1 | |||
| AK291258.1 | ||||
| AK299179.1 | ||||
| AK300284.1 | ||||
| AK307730.1 | ||||
| BC039060.1 | AAH39060.1 | |||
| BC040540.1 | AAH40540.1 | |||
| L41870.1 | AAB59465.1 | |||
| M15400.1 | AAA69807.1 | |||
| M28419.1 | AAA69808.1 | |||
| M33647.1 | AAA69806.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv832602 | Chr.13:48429023 - 48593645 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs116357023] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map