Assay Details
Target Gene Details
Entrez Gene ID: | 255967 |
Gene Name: | PAN3 poly(A) specific ribonuclease subunit |
Gene Aliases: |
- |
Location: |
Chr.13:28138079-28295338 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PAN3 | NM_175854.7 | NP_787050.6 | ||
| XM_005266332.3 | XP_005266389.1 | |||
| XM_005266333.3 | XP_005266390.1 | |||
| XM_005266334.1 | XP_005266391.1 | |||
| XM_011535032.2 | XP_011533334.1 | |||
| XM_011535033.2 | XP_011533335.1 | |||
| XM_011535034.2 | XP_011533336.1 | |||
| XM_017020528.1 | XP_016876017.1 | |||
| XM_017020529.1 | XP_016876018.1 | |||
| XM_017020530.1 | XP_016876019.1 | |||
| XM_017020532.1 | XP_016876021.1 | |||
| XM_017020533.1 | XP_016876022.1 | |||
| AB107584.1 | BAD02262.1 | |||
| AB109552.1 | BAD93184.1 | |||
| AK296435.1 | ||||
| BC128179.1 | ||||
| BC128180.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv34052 | Chr.13:28074532 - 28352440 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map